Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BECN1
Entrez Gene ID
8678
Gene Name
beclin 1
Gene Aliases
ATG6, VPS30, beclin1
Chromosome Location
Chr.17: 42810132 - 42824316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313998.1 NP_001300927.1 11 - 12 1341 66
NM_003766.4 NP_003757.1 11 - 12 1363 66
GenBank mRNA AF077301.1 - 10 - 11 1177 66
AF139131.1 - 11 - 12 1297 66
AK223411.1 - 11 - 12 1305 66
AK299616.1 - 10 - 11 1985 66
AK312651.1 - 11 - 12 1288 66
BC010276.1 - 11 - 12 1310 66
BX647788.1 - 8 - 9 4066 66
L38932.1 - 6 - 7 732 66

View Full Product Details