Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BECN1
Entrez Gene ID
8678
Gene Name
beclin 1
Gene Aliases
ATG6, VPS30, beclin1
Chromosome Location
Chr.17: 42810132 - 42824316 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313998.1 NP_001300927.1 8 - 9 994 87
NM_001313999.1 NP_001300928.1 7 - 8 832 87
NM_001314000.1 NP_001300929.1 6 - 7 766 87
NM_003766.4 NP_003757.1 8 - 9 1016 87
XM_005257759.2 XP_005257816.1 8 - 9 938 87
XM_005257760.3 XP_005257817.1 8 - 9 938 87
XM_011525421.2 XP_011523723.1 8 - 9 938 87
XM_017025262.1 XP_016880751.1 8 - 9 1011 87
XM_017025263.1 XP_016880752.1 8 - 9 1011 87
XM_017025264.1 XP_016880753.1 8 - 9 1011 87
GenBank mRNA AF077301.1 - 7 - 8 830 87
AF139131.1 - 8 - 9 950 87
AK223411.1 - 8 - 9 958 87
AK298619.1 - 6 - 7 738 87
AK299616.1 - 7 - 8 1638 87
AK312651.1 - 8 - 9 941 87
BC010276.1 - 8 - 9 963 87
BX647788.1 - 6 - 7 1733 87
KC776730.1 - 7 - 8 830 87
L38932.1 - 3 - 4 385 87

View Full Product Details