The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175066.3 | NP_778236.2 | 13 - 14 | 1874 | 82 |
GenBank mRNA | AB593157.1 | - | 13 - 14 | 1894 | 82 |
AK097078.1 | - | 13 - 14 | 1874 | 82 | |
BC012461.2 | - | 4 - 5 | 397 | 82 | |
BC040185.2 | - | 13 - 14 | 1847 | 82 |