The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258006.1 | NP_001244935.1 | 3 - 4 | 449 | 105 |
NM_016129.2 | NP_057213.2 | 3 - 4 | 449 | 105 | |
XM_011532019.1 | XP_011530321.1 | 3 - 4 | 445 | 105 | |
GenBank mRNA | AF100757.1 | - | 3 - 4 | 313 | 105 |
AK001148.1 | - | 3 - 4 | 445 | 105 | |
AK024005.1 | - | 3 - 4 | 334 | 105 | |
AK094238.1 | - | 3 - 4 | 1087 | 105 | |
AK223178.1 | - | 3 - 4 | 319 | 105 | |
BC004302.1 | - | 3 - 4 | 312 | 105 | |
BC009292.2 | - | 3 - 4 | 312 | 105 | |
BC093007.1 | - | 3 - 4 | 334 | 105 | |
KU178633.1 | - | 3 - 4 | 310 | 105 | |
KU178634.1 | - | 3 - 4 | 310 | 105 |