The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258006.1 | NP_001244935.1 | 6 - 7 | 853 | 86 |
NM_016129.2 | NP_057213.2 | 6 - 7 | 853 | 86 | |
XM_011532019.1 | XP_011530321.1 | 6 - 7 | 849 | 86 | |
GenBank mRNA | AF100757.1 | - | 6 - 7 | 717 | 86 |
AK001148.1 | - | 6 - 7 | 849 | 86 | |
AK001210.1 | - | 6 - 7 | 680 | 86 | |
AK024005.1 | - | 6 - 7 | 738 | 86 | |
AK094238.1 | - | 6 - 7 | 1491 | 86 | |
AK223178.1 | - | 6 - 7 | 723 | 86 | |
BC004302.1 | - | 6 - 7 | 716 | 86 | |
BC009292.2 | - | 6 - 7 | 716 | 86 | |
BC093007.1 | - | 6 - 7 | 738 | 86 | |
KU178633.1 | - | 6 - 7 | 714 | 86 | |
KU178634.1 | - | 6 - 7 | 714 | 86 |