The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016129.2 | NP_057213.2 | 8 - 9 | 1138 | 147 |
XM_011532019.1 | XP_011530321.1 | 8 - 9 | 1134 | 147 | |
GenBank mRNA | AF100757.1 | - | 8 - 9 | 1002 | 147 |
AK001148.1 | - | 8 - 9 | 147 | ||
AK001210.1 | - | 8 - 9 | 965 | 147 | |
AK024005.1 | - | 8 - 9 | 1023 | 147 | |
AK223178.1 | - | 8 - 9 | 1008 | 147 | |
BC004302.1 | - | 8 - 9 | 1001 | 147 | |
BC009292.2 | - | 8 - 9 | 1001 | 147 | |
BC093007.1 | - | 8 - 9 | 1023 | 147 | |
KU178633.1 | - | 8 - 9 | 999 | 147 | |
KU178634.1 | - | 8 - 9 | 999 | 147 |