The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317231.1 | NP_001304160.1 | 5 - 6 | 551 | 93 |
NM_001317232.1 | NP_001304161.1 | 4 - 5 | 524 | 93 | |
NM_014342.3 | NP_055157.1 | 5 - 6 | 551 | 93 | |
XM_006718172.2 | XP_006718235.1 | 5 - 6 | 494 | 93 | |
XM_011519959.2 | XP_011518261.1 | 5 - 6 | 494 | 93 | |
XM_011519960.2 | XP_011518262.1 | 4 - 5 | 514 | 93 | |
XM_011519961.2 | XP_011518263.1 | 5 - 6 | 494 | 93 | |
XM_017017462.1 | XP_016872951.1 | 4 - 5 | 514 | 93 | |
XM_017017463.1 | XP_016872952.1 | 5 - 6 | 470 | 93 | |
GenBank mRNA | AF085361.1 | - | 5 - 6 | 410 | 93 |
AF176008.1 | - | 5 - 6 | 411 | 93 | |
AK223097.1 | - | 5 - 6 | 374 | 93 | |
AK313032.1 | - | 5 - 6 | 494 | 93 | |
AY380792.1 | - | 5 - 6 | 409 | 93 | |
BC000875.1 | - | 5 - 6 | 407 | 93 | |
BQ960010.1 | - | 4 - 5 | 396 | 93 |