The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317231.1 | NP_001304160.1 | 8 - 9 | 729 | 72 |
NM_001317232.1 | NP_001304161.1 | 7 - 8 | 702 | 72 | |
NM_001317233.1 | NP_001304162.1 | 7 - 8 | 671 | 72 | |
NM_014342.3 | NP_055157.1 | 8 - 9 | 729 | 72 | |
XM_006718172.2 | XP_006718235.1 | 8 - 9 | 672 | 72 | |
XM_011519959.2 | XP_011518261.1 | 8 - 9 | 672 | 72 | |
XM_011519960.2 | XP_011518262.1 | 7 - 8 | 692 | 72 | |
XM_011519961.2 | XP_011518263.1 | 8 - 9 | 672 | 72 | |
XM_017017462.1 | XP_016872951.1 | 7 - 8 | 692 | 72 | |
XM_017017463.1 | XP_016872952.1 | 8 - 9 | 648 | 72 | |
GenBank mRNA | AF085361.1 | - | 8 - 9 | 588 | 72 |
AF176008.1 | - | 8 - 9 | 589 | 72 | |
AK223097.1 | - | 8 - 9 | 552 | 72 | |
AK295218.1 | - | 7 - 8 | 551 | 72 | |
AK313032.1 | - | 8 - 9 | 672 | 72 | |
AY380792.1 | - | 8 - 9 | 587 | 72 | |
BC000875.1 | - | 8 - 9 | 585 | 72 | |
BQ960010.1 | - | 7 - 8 | 574 | 72 |