Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MUTYH
- Entrez Gene ID
-
4595
- Gene Name
- mutY DNA glycosylase
- Gene Aliases
-
MYH
- Chromosome Location
-
Chr.1: 45329242 - 45340925 on Build GRCh38
View Full Product Details