Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GUCY1A3
Entrez Gene ID
2982
Gene Name
guanylate cyclase 1 soluble subunit alpha
Gene Aliases
GC-SA3, GUC1A3, GUCA3, GUCSA3, GUCY1A1, MYMY6
Chromosome Location
Chr.4: 155666710 - 155737062 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000856.5 NP_000847.2 3 - 4 422 134
GenBank mRNA Y15723.1 - 3 - 4 409 134

View Full Product Details