The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000856.5 | NP_000847.2 | 7 - 8 | 1620 | 110 |
NM_001130682.2 | NP_001124154.1 | 6 - 7 | 1526 | 110 | |
NM_001130683.3 | NP_001124155.1 | 6 - 7 | 1494 | 110 | |
NM_001130684.2 | NP_001124156.1 | 5 - 6 | 1466 | 110 | |
NM_001130685.2 | NP_001124157.1 | 6 - 7 | 1315 | 110 | |
NM_001130687.2 | NP_001124159.1 | 6 - 7 | 1526 | 110 | |
NM_001256449.1 | NP_001243378.1 | 6 - 7 | 1457 | 110 | |
XM_005262955.2 | XP_005263012.1 | 6 - 7 | 1488 | 110 | |
XM_005262956.2 | XP_005263013.1 | 7 - 8 | 1303 | 110 | |
XM_005262957.2 | XP_005263014.1 | 7 - 8 | 1408 | 110 | |
XM_006714196.2 | XP_006714259.1 | 6 - 7 | 1475 | 110 | |
XM_006714197.2 | XP_006714260.1 | 6 - 7 | 1250 | 110 | |
XM_006714198.2 | XP_006714261.1 | 6 - 7 | 1135 | 110 | |
XM_011531900.2 | XP_011530202.1 | 6 - 7 | 1214 | 110 |