Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HDGFRP3
Entrez Gene ID
50810
Gene Name
hepatoma-derived growth factor, related protein 3
Gene Aliases
CGI-142, HDGF-2, HDGF2, HRP-3
Chromosome Location
Chr.15: 83112738 - 83208018 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016073.3 NP_057157.1 1 - 2 693 96
XM_006720554.3 XP_006720617.1 1 - 2 229 96
GenBank mRNA AB029156.1 - 1 - 2 209 96
AF110642.1 - 1 - 2 213 96
AF151900.1 - 1 - 2 213 96
AK001280.1 - 1 - 2 244 96
BC015483.1 - 1 - 2 187 96
BC040554.1 - 1 - 2 696 96

View Full Product Details