The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016073.3 | NP_057157.1 | 4 - 5 | 1058 | 128 |
XM_006720554.3 | XP_006720617.1 | 4 - 5 | 594 | 128 | |
GenBank mRNA | AB029156.1 | - | 4 - 5 | 574 | 128 |
AF110642.1 | - | 4 - 5 | 578 | 128 | |
AF151900.1 | - | 4 - 5 | 578 | 128 | |
AK001280.1 | - | 4 - 5 | 609 | 128 | |
BC015483.1 | - | 4 - 5 | 552 | 128 | |
BC040554.1 | - | 4 - 5 | 1061 | 128 |