The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005131.2 | NP_005122.2 | 16 - 17 | 1347 | 87 |
XM_011525772.2 | XP_011524074.1 | 16 - 17 | 1368 | 87 | |
XM_011525773.1 | XP_011524075.1 | 17 - 18 | 1398 | 87 | |
XM_011525774.2 | XP_011524076.1 | 6 - 7 | 1939 | 87 | |
XM_017026103.1 | XP_016881592.1 | 13 - 14 | 2888 | 87 | |
GenBank mRNA | AK055354.1 | - | 5 - 6 | 2018 | 87 |
AK123530.1 | - | 3 - 4 | 1484 | 87 | |
AK225067.1 | - | 16 - 17 | 1323 | 87 | |
AK225709.1 | - | 16 - 17 | 1311 | 87 | |
AK314755.1 | - | 16 - 17 | 1313 | 87 | |
BC010381.1 | - | 16 - 17 | 1338 | 87 | |
L36529.1 | - | 16 - 17 | 87 |