Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THOC1
Entrez Gene ID
9984
Gene Name
THO complex 1
Gene Aliases
HPR1, P84, P84N5
Chromosome Location
Chr.18: 214348 - 268059 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005131.2 NP_005122.2 6 - 7 459 171
XM_011525772.2 XP_011524074.1 6 - 7 459 171
XM_011525773.1 XP_011524075.1 7 - 8 489 171
XM_017026103.1 XP_016881592.1 3 - 4 1979 171
XM_017026104.1 XP_016881593.1 6 - 7 459 171
GenBank mRNA AK225067.1 - 6 - 7 435 171
AK225709.1 - 6 - 7 423 171
AK314755.1 - 6 - 7 425 171
AY573303.1 - 6 - 7 419 171
BC010381.1 - 6 - 7 450 171
L36529.1 - 6 - 7 433 171

View Full Product Details