The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033044.3 | NP_001028216.1 | 1 - 2 | 528 | 115 |
GenBank mRNA | AK122784.1 | - | 1 - 2 | 528 | 115 |
BC010037.2 | - | 1 - 2 | 100 | 115 | |
BC011700.2 | - | 1 - 2 | 96 | 115 | |
BC011852.2 | - | 1 - 2 | 107 | 115 | |
BG771732.1 | - | 1 - 2 | 147 | 116 | |
BX537384.1 | - | 1 - 2 | 112 | 115 | |
DA151535.1 | - | 1 - 2 | 173 | 115 | |
Y00387.2 | - | 1 - 2 | 106 | 114 |