Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- H2AFY
- Entrez Gene ID
-
9555
- Gene Name
- H2A histone family member Y
- Gene Aliases
-
H2A.y, H2A/y, H2AF12M, MACROH2A1.1, mH2A1, macroH2A1.2
- Chromosome Location
-
Chr.5: 135334381 - 135399887 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK023833.1 |
- |
1 - 1 |
5764 |
113 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AU158717.1 |
- |
1 |
|
|
Chr. 5 - 135351582 - 135352080 [+] on Build GRCh38 |
113 |
GenBankmRNA |
DB358346.1 |
- |
1 |
|
|
Chr. 5 - 135351744 - 135352294 [+] on Build GRCh38 |
114 |
GenBankmRNA |
BQ334796.1 |
- |
1 |
|
|
Chr. 5 - 135351823 - 135352085 [+] on Build GRCh38 |
114 |
GenBankmRNA |
AW080273.1 |
- |
1 |
|
|
Chr. 5 - 135351744 - 135352268 [+] on Build GRCh38 |
113 |
GenBankmRNA |
DA463499.1 |
- |
1 |
413 |
|
Chr. 5 - 135351808 - 135352363 [-] on Build GRCh38 |
113 |
GenBankmRNA |
BF901854.1 |
- |
1 |
|
|
Chr. 5 - 135351798 - 135352124 [+] on Build GRCh38 |
113 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
chromosome 5 open reading frame 66 |
C5orf66 |
|
-7.1kb
|
Sense |
Chr. - 135033280 - 135344680 on Build GRCh38 |
Downstream |
dendritic cell associated nuclear protein |
DCANP1 |
C5orf20, DCNP1 |
91.9kb
|
Anti-sense |
Chr. - 135444214 - 135447348 on Build GRCh38 |
View Full Product Details