The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270660.1 | NP_001257589.1 | 14 - 15 | 1448 | 88 |
NM_001270661.1 | NP_001257590.1 | 12 - 13 | 1277 | 88 | |
NM_005885.3 | NP_005876.2 | 15 - 16 | 1592 | 88 | |
XM_011513932.2 | XP_011512234.1 | 15 - 16 | 1350 | 88 | |
XM_011513934.1 | XP_011512236.1 | 15 - 16 | 1553 | 88 | |
XM_011513936.2 | XP_011512238.1 | 15 - 16 | 1553 | 88 | |
XM_011513937.2 | XP_011512239.1 | 5 - 6 | 934 | 88 | |
GenBank mRNA | AB011169.1 | - | 15 - 16 | 1515 | 88 |
AF009301.1 | - | 2 - 3 | 211 | 88 | |
AK296585.1 | - | 12 - 13 | 1200 | 88 | |
AK300034.1 | - | 14 - 15 | 1384 | 88 | |
AK314686.1 | - | 4 - 5 | 445 | 88 | |
BC046148.1 | - | 7 - 8 | 509 | 88 | |
BC136461.1 | - | 15 - 16 | 1464 | 88 | |
BC136462.1 | - | 15 - 16 | 1464 | 88 | |
BC142679.1 | - | 15 - 16 | 1443 | 88 | |
BC142694.1 | - | 15 - 16 | 1443 | 88 |