Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCH6
Entrez Gene ID
10299
Gene Name
membrane associated ring-CH-type finger 6
Gene Aliases
DOA10, MARCH-VI, RNF176, TEB4
Chromosome Location
Chr.5: 10353639 - 10440388 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270660.1 NP_001257589.1 22 - 23 2489 97
NM_001270661.1 NP_001257590.1 20 - 21 2318 97
NM_005885.3 NP_005876.2 23 - 24 2633 97
XM_011513932.2 XP_011512234.1 23 - 24 2391 97
XM_011513937.2 XP_011512239.1 13 - 14 1975 97
GenBank mRNA AB011169.1 - 23 - 24 2556 97
AF009301.1 - 10 - 11 1252 97
AK296585.1 - 20 - 21 2241 97
AK300034.1 - 22 - 23 2425 97
AK314686.1 - 12 - 13 1486 97
BC046148.1 - 15 - 16 1550 97
BC136461.1 - 23 - 24 2505 97
BC136462.1 - 23 - 24 2505 97
BC142679.1 - 23 - 24 2484 97
BC142694.1 - 23 - 24 2484 97

View Full Product Details