The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000884.2 | NP_000875.2 | 13 - 14 | 1620 | 128 |
XM_006713128.3 | XP_006713191.1 | 14 - 15 | 2000 | 128 | |
XM_017006349.1 | XP_016861838.1 | 13 - 14 | 1877 | 128 | |
XM_017006350.1 | XP_016861839.1 | 12 - 13 | 1805 | 128 | |
GenBank mRNA | BC006124.1 | - | 13 - 14 | 1568 | 128 |
BC012840.1 | - | 13 - 14 | 1561 | 128 | |
BC015567.2 | - | 13 - 14 | 1559 | 128 | |
J04208.1 | - | 13 - 14 | 1575 | 128 |