Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABPA
Entrez Gene ID
2551
Gene Name
GA binding protein transcription factor alpha subunit
Gene Aliases
E4TF1-60, E4TF1A, NFT2, NRF2, NRF2A, RCH04A07
Chromosome Location
Chr.21: 25734947 - 25772460 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197297.1 NP_001184226.1 6 - 7 902 105
NM_002040.3 NP_002031.2 6 - 7 1339 105
XM_005260938.4 XP_005260995.1 6 - 7 974 105
XM_011529520.2 XP_011527822.1 7 - 8 1378 105
XM_011529521.2 XP_011527823.1 8 - 9 1469 105
XM_017028313.1 XP_016883802.1 6 - 7 985 105
GenBank mRNA AK223164.1 - 6 - 7 941 105
AK313545.1 - 6 - 7 1136 105
BC035031.2 - 6 - 7 972 105
BX647755.1 - 6 - 7 1280 105
D13318.1 - 6 - 7 944 105
EU159453.1 - 5 - 6 761 105
JQ313900.1 - 5 - 6 752 105
U13044.1 - 6 - 7 937 105

View Full Product Details