The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256310.1 | NP_001243239.1 | 9 - 10 | 1394 | 207 |
NM_014905.4 | NP_055720.3 | 9 - 10 | 1394 | 207 | |
XM_005246467.3 | XP_005246524.1 | 9 - 10 | 1394 | 207 | |
XM_006712435.1 | XP_006712498.1 | 9 - 10 | 1394 | 207 | |
GenBank mRNA | AB020645.1 | - | 9 - 10 | 1380 | 207 |
AF097492.1 | - | 9 - 10 | 1028 | 207 | |
AF158555.1 | - | 10 - 11 | 1374 | 207 | |
AF223943.1 | - | 9 - 10 | 1149 | 207 | |
AF279697.1 | - | 6 - 7 | 479 | 207 | |
AK001220.1 | - | 6 - 7 | 466 | 207 | |
AK289747.1 | - | 10 - 11 | 1370 | 207 | |
BC038507.1 | - | 10 - 11 | 1370 | 207 |