The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282400.1 | NP_001269329.1 | 7 - 8 | 1311 | 154 |
GenBank mRNA | AK094141.1 | - | 7 - 8 | 1272 | 154 |