Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WWC3
Entrez Gene ID
55841
Gene Name
WWC family member 3
Gene Aliases
BM042
Chromosome Location
Chr.X: 10015755 - 10144478 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015691.3 NP_056506.2 15 - 16 2305 123
GenBank mRNA AB033106.1 - 13 - 14 1928 123
AK024683.1 - 6 - 7 1115 123
AK091936.1 - 6 - 7 982 123
AK292961.1 - 14 - 15 2187 123
AL353937.1 - 12 - 13 1796 123
BC032447.1 - 12 - 13 1840 123
KC987947.1 - 15 - 16 2482 123

View Full Product Details