Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCGR
Entrez Gene ID
2642
Gene Name
glucagon receptor
Gene Aliases
GGR, GL-R
Chromosome Location
Chr.17: 81804078 - 81814013 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000160.4 NP_000151.1 11 - 12 1335 69
XM_006722277.1 XP_006722340.1 11 - 12 1364 69
XM_011523539.1 XP_011521841.1 11 - 12 1462 69
XM_017024446.1 XP_016879935.1 11 - 12 1383 69
XM_017024447.1 XP_016879936.1 10 - 11 1172 69
GenBank mRNA BC104854.1 - 10 - 11 1085 69
BC112041.1 - 10 - 11 1084 69
L20316.1 - 11 - 12 1317 69
U03469.1 - 10 - 11 1101 69

View Full Product Details