The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004932.3 | NP_004923.1 | 2 - 3 | 587 | 106 |
XM_011513921.2 | XP_011512223.1 | 2 - 3 | 406 | 106 | |
XM_017008910.1 | XP_016864399.1 | 1 - 2 | 357 | 106 | |
XM_017008911.1 | XP_016864400.1 | 2 - 3 | 544 | 106 | |
GenBank mRNA | AK291290.1 | - | 2 - 3 | 553 | 106 |
BC000019.2 | - | 2 - 3 | 492 | 106 | |
D31784.1 | - | 1 - 2 | 347 | 106 |