Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
Chromosome Location
Chr.2: 25232961 - 25342590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175629.2 NP_783328.1 1 - 2 161 75
NM_175630.1 NP_783329.1 1 - 2 161 75
NR_135490.1 1 - 2 161 75
GenBank mRNA AB208833.1 - 1 - 2 60 75
AF331856.1 - 1 - 2 40 75
BC032392.1 - 1 - 2 161 75
BC051864.1 - 1 - 2 29 75

View Full Product Details