Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
Chromosome Location
Chr.2: 25232961 - 25342590 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320893.1 NP_001307822.1 4 - 5 704 72
NM_022552.4 NP_072046.2 9 - 10 1390 72
NM_153759.3 NP_715640.2 5 - 6 674 72
NM_175629.2 NP_783328.1 9 - 10 1461 72
NR_135490.1 9 - 10 1461 72
XM_005264175.4 XP_005264232.1 9 - 10 1324 72
XM_005264177.4 XP_005264234.1 4 - 5 616 72
XM_011532662.2 XP_011530964.1 7 - 8 1326 72
XM_011532663.2 XP_011530965.1 10 - 11 1503 72
XM_011532664.2 XP_011530966.1 9 - 10 1401 72
XM_011532666.2 XP_011530968.1 5 - 6 689 72
XM_011532667.2 XP_011530969.1 4 - 5 506 72
XM_017003526.1 XP_016859015.1 9 - 10 1432 72
XM_017003527.1 XP_016859016.1 4 - 5 660 72
GenBank mRNA AB208833.1 - 9 - 10 1360 72
AF067972.2 - 9 - 10 1352 72
AF331856.1 - 9 - 10 1340 72
AF480163.1 - 5 - 6 670 72
BC043617.1 - 9 - 10 1360 72
BM470515.1 - - 686 72

View Full Product Details