Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAGI1
Entrez Gene ID
9223
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 1
Gene Aliases
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
Chromosome Location
Chr.3: 65353525 - 66038836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004742.2 NP_004733.2 14 - 15 2947 74
NM_015520.1 NP_056335.1 14 - 15 2947 74
XM_005265563.1 XP_005265620.1 14 - 15 2955 74
XM_005265564.1 XP_005265621.1 14 - 15 2949 74
XM_005265565.1 XP_005265622.1 14 - 15 2955 74
XM_005265566.1 XP_005265623.1 14 - 15 2949 74
XM_005265568.1 XP_005265625.1 13 - 14 2913 74
XM_005265570.1 XP_005265627.1 14 - 15 2955 74
XM_005265574.3 XP_005265631.1 14 - 15 2949 74
XM_006713407.1 XP_006713470.1 14 - 15 2955 74
XM_006713408.1 XP_006713471.1 14 - 15 2952 74
XM_006713409.1 XP_006713472.1 14 - 15 2952 74
XM_006713410.1 XP_006713473.1 13 - 14 2913 74
XM_006713414.3 XP_006713477.1 14 - 15 2955 74
XM_011534236.1 XP_011532538.1 13 - 14 2916 74
XM_011534240.2 XP_011532542.1 2 - 3 4311 74
XM_017007497.1 XP_016862986.1 14 - 15 2952 74
XM_017007498.1 XP_016862987.1 14 - 15 2949 74
XM_017007499.1 XP_016862988.1 14 - 15 3310 74
XM_017007500.1 XP_016862989.1 14 - 15 3303 74
XM_017007502.1 XP_016862991.1 14 - 15 2952 74
XM_017007503.1 XP_016862992.1 14 - 15 2955 74
XM_017007504.1 XP_016862993.1 14 - 15 2952 74
XM_017007505.1 XP_016862994.1 14 - 15 2952 74
XM_017007506.1 XP_016862995.1 14 - 15 2949 74
GenBank mRNA AB010894.1 - 14 - 15 2663 74
AF401654.1 - 14 - 15 2421 74
AK289803.1 - 14 - 15 2516 74
AK307794.1 - 2 - 3 397 74
U80754.1 - 12 - 13 1974 74

View Full Product Details