Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAGI1
Entrez Gene ID
9223
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 1
Gene Aliases
AIP-3, AIP3, BAIAP1, BAP-1, BAP1, MAGI-1, Magi1d, TNRC19, WWP3
Chromosome Location
Chr.3: 65353525 - 66038836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033057.1 NP_001028229.1 18 - 19 3723 62
NM_004742.2 NP_004733.2 19 - 20 3810 62
NM_015520.1 NP_056335.1 19 - 20 3807 62
XM_005265563.1 XP_005265620.1 19 - 20 3815 62
XM_005265564.1 XP_005265621.1 19 - 20 3812 62
XM_005265565.1 XP_005265622.1 19 - 20 3782 62
XM_005265566.1 XP_005265623.1 19 - 20 3776 62
XM_005265568.1 XP_005265625.1 18 - 19 3773 62
XM_005265574.3 XP_005265631.1 19 - 20 3812 62
XM_006713407.1 XP_006713470.1 19 - 20 3818 62
XM_006713408.1 XP_006713471.1 19 - 20 3815 62
XM_006713409.1 XP_006713472.1 19 - 20 3815 62
XM_006713410.1 XP_006713473.1 18 - 19 3776 62
XM_006713411.1 XP_006713474.1 18 - 19 3734 62
XM_006713412.1 XP_006713475.1 18 - 19 3728 62
XM_006713414.3 XP_006713477.1 19 - 20 3818 62
XM_011534236.1 XP_011532538.1 18 - 19 3779 62
XM_011534240.2 XP_011532542.1 7 - 8 5174 62
XM_011534241.2 XP_011532543.1 6 - 7 5088 62
XM_017007497.1 XP_016862986.1 19 - 20 3812 62
XM_017007498.1 XP_016862987.1 19 - 20 3809 62
XM_017007499.1 XP_016862988.1 19 - 20 4173 62
XM_017007500.1 XP_016862989.1 19 - 20 4163 62
XM_017007501.1 XP_016862990.1 18 - 19 3695 62
XM_017007502.1 XP_016862991.1 19 - 20 3815 62
XM_017007503.1 XP_016862992.1 19 - 20 3815 62
XM_017007504.1 XP_016862993.1 19 - 20 3812 62
XM_017007505.1 XP_016862994.1 19 - 20 3812 62
XM_017007506.1 XP_016862995.1 19 - 20 3776 62
XM_017007507.1 XP_016862996.1 17 - 18 2856 62
GenBank mRNA AB010894.1 - 19 - 20 3526 62
AF401654.1 - 19 - 20 3281 62
AF401656.1 - 18 - 19 3197 62
AK289803.1 - 19 - 20 3379 62

View Full Product Details