Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLCO2B1
Entrez Gene ID
11309
Gene Name
solute carrier organic anion transporter family member 2B1
Gene Aliases
OATP-B, OATP2B1, OATPB, SLC21A9
Chromosome Location
Chr.11: 75150987 - 75206400 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145211.2 NP_001138683.1 13 - 14 2039 94
NM_001145212.2 NP_001138684.1 10 - 11 1912 94
NM_007256.4 NP_009187.1 13 - 14 2344 94
XM_017017157.1 XP_016872646.1 13 - 14 1962 94
GenBank mRNA AB020687.1 - 13 - 14 2215 94
AB026256.1 - 13 - 14 2127 94
AF205073.1 - 13 - 14 2061 94
AK127289.1 - 1 - 2 3874 94
AK127363.1 - 1 - 2 1155 94
AK290234.1 - 13 - 14 2226 94
AK294503.1 - 13 - 14 2024 94
AK296079.1 - 4 - 5 537 94
AK300134.1 - 10 - 11 1784 94
AK302492.1 - 11 - 12 1989 94
AL117465.1 - 9 - 10 1516 94
BC041095.1 - 13 - 14 2213 94

View Full Product Details