The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145211.2 | NP_001138683.1 | 5 - 6 | 771 | 70 |
NM_001145212.2 | NP_001138684.1 | 2 - 3 | 644 | 70 | |
NM_007256.4 | NP_009187.1 | 5 - 6 | 1076 | 70 | |
XM_017017157.1 | XP_016872646.1 | 5 - 6 | 694 | 70 | |
GenBank mRNA | AB020687.1 | - | 5 - 6 | 947 | 70 |
AB026256.1 | - | 5 - 6 | 859 | 70 | |
AK290234.1 | - | 5 - 6 | 958 | 70 | |
AK294503.1 | - | 5 - 6 | 756 | 70 | |
AK300134.1 | - | 2 - 3 | 516 | 70 | |
AK302492.1 | - | 3 - 4 | 721 | 70 | |
BC041095.1 | - | 5 - 6 | 945 | 70 |