Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARHGEF7
Entrez Gene ID
8874
Gene Name
Rho guanine nucleotide exchange factor 7
Gene Aliases
BETA-PIX, COOL-1, COOL1, Nbla10314, P50, P50BP, P85, P85COOL1, P85SPR, PAK3, PIXB
Chromosome Location
Chr.13: 111114619 - 111305734 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113511.2 NP_001106983.1 5 - 6 1439 95
NM_001113512.2 NP_001106984.1 3 - 4 1289 95
NM_001113513.1 NP_001106985.1 3 - 4 381 95
NM_001320852.1 NP_001307781.1 4 - 5 1376 95
NM_001320853.1 NP_001307782.1 3 - 4 287 95
NM_003899.3 NP_003890.1 3 - 4 494 95
NM_145735.3 NP_663788.1 4 - 5 1376 95
XM_006719956.3 XP_006720019.1 4 - 5 504 95
XM_011521133.2 XP_011519435.1 4 - 5 504 95
XM_017020814.1 XP_016876303.1 4 - 5 504 95
XM_017020816.1 XP_016876305.1 3 - 4 272 95
XM_017020818.1 XP_016876307.1 4 - 5 372 95
XM_017020819.1 XP_016876308.1 3 - 4 381 95
XM_017020820.1 XP_016876309.1 3 - 4 504 95
XM_017020824.1 XP_016876313.1 4 - 5 372 95
XM_017020826.1 XP_016876315.1 4 - 5 372 95
GenBank mRNA AB177848.1 - 4 - 5 1377 95
AK300272.1 - 3 - 4 274 95
AL834228.1 - 4 - 5 560 95
BC033905.1 - 3 - 4 486 95
BC050521.1 - 3 - 4 353 95
BC060776.1 - 5 - 6 781 95
D63476.1 - 3 - 4 95
DA404477.1 - 4 - 5 95
DA439385.1 - 5 - 6 459 95
DC318118.1 - 3 - 4 287 95

View Full Product Details