The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042355.1 | NP_001035814.1 | 11 - 12 | 2224 | 105 |
NM_001042412.1 | NP_001035877.1 | 11 - 12 | 2248 | 105 | |
NM_015247.2 | NP_056062.1 | 13 - 14 | 2370 | 105 | |
XM_005255812.2 | XP_005255869.1 | 12 - 13 | 2321 | 105 | |
XM_006721149.1 | XP_006721212.1 | 12 - 13 | 2385 | 105 | |
XM_011522907.2 | XP_011521209.1 | 11 - 12 | 2147 | 105 | |
XM_011522908.1 | XP_011521210.1 | 11 - 12 | 2209 | 105 | |
XM_017022977.1 | XP_016878466.1 | 11 - 12 | 2519 | 105 | |
XM_017022978.1 | XP_016878467.1 | 11 - 12 | 2315 | 105 | |
XM_017022979.1 | XP_016878468.1 | 11 - 12 | 2275 | 105 | |
XM_017022980.1 | XP_016878469.1 | 11 - 12 | 2308 | 105 | |
XM_017022981.1 | XP_016878470.1 | 11 - 12 | 2176 | 105 | |
GenBank mRNA | AF161542.1 | - | 10 - 11 | 2196 | 105 |
AJ250014.1 | - | 13 - 14 | 2346 | 105 | |
AK000187.1 | - | 6 - 7 | 1026 | 105 | |
AK024212.1 | - | 9 - 10 | 1682 | 105 | |
AK024348.1 | - | 9 - 10 | 1731 | 105 | |
AK310832.1 | - | 11 - 12 | 2196 | 105 | |
BC012342.1 | - | 11 - 12 | 2222 | 105 |