The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042355.1 | NP_001035814.1 | 4 - 5 | 1079 | 73 |
NM_001042412.1 | NP_001035877.1 | 4 - 5 | 1103 | 73 | |
NM_015247.2 | NP_056062.1 | 5 - 6 | 1216 | 73 | |
XM_005255812.2 | XP_005255869.1 | 5 - 6 | 1176 | 73 | |
XM_006721149.1 | XP_006721212.1 | 5 - 6 | 1240 | 73 | |
XM_011522907.2 | XP_011521209.1 | 4 - 5 | 1002 | 73 | |
XM_011522908.1 | XP_011521210.1 | 4 - 5 | 1094 | 73 | |
XM_011522911.2 | XP_011521213.1 | 4 - 5 | 1094 | 73 | |
XM_017022977.1 | XP_016878466.1 | 4 - 5 | 1374 | 73 | |
XM_017022978.1 | XP_016878467.1 | 4 - 5 | 1170 | 73 | |
XM_017022979.1 | XP_016878468.1 | 4 - 5 | 1130 | 73 | |
XM_017022980.1 | XP_016878469.1 | 4 - 5 | 1163 | 73 | |
XM_017022981.1 | XP_016878470.1 | 4 - 5 | 1061 | 73 | |
GenBank mRNA | AJ250014.1 | - | 5 - 6 | 1192 | 73 |
AK024212.1 | - | 2 - 3 | 537 | 73 | |
AK024348.1 | - | 2 - 3 | 586 | 73 | |
AK056226.1 | - | 4 - 5 | 1034 | 73 | |
AK225202.1 | - | 5 - 6 | 1196 | 73 | |
AK292975.1 | - | 4 - 5 | 1061 | 73 | |
AK310832.1 | - | 4 - 5 | 1051 | 73 | |
BC012342.1 | - | 4 - 5 | 1077 | 73 |