The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015510.4 | NP_056325.2 | 2 - 3 | 243 | 76 |
XM_005256595.4 | XP_005256652.2 | 3 - 4 | 1037 | 76 | |
XM_005256596.3 | XP_005256653.1 | 2 - 3 | 497 | 76 | |
XM_011523786.2 | XP_011522088.1 | 3 - 4 | 1037 | 76 | |
XM_017024425.1 | XP_016879914.1 | 2 - 3 | 355 | 76 | |
XM_017024426.1 | XP_016879915.1 | 3 - 4 | 582 | 76 | |
GenBank mRNA | AF151851.1 | - | 2 - 3 | 227 | 76 |
AL117567.1 | - | 2 - 3 | 230 | 76 | |
AY358498.2 | - | 1 - 2 | 191 | 76 | |
BC004126.2 | - | 2 - 3 | 217 | 76 | |
BC009679.2 | - | 2 - 3 | 216 | 76 | |
BI460496.1 | - | 2 - 3 | 244 | 76 | |
CR457124.1 | - | 2 - 3 | 199 | 76 |