The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293163.1 | NP_001280092.1 | 11 - 12 | 1539 | 61 |
GenBank mRNA | BC016925.1 | - | 11 - 12 | 1485 | 61 |
EU159452.1 | - | 10 - 11 | 1408 | 61 |