Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
Chromosome Location
Chr.X: 44146254 - 44343677 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025184.3 NP_079460.2 12 - 13 2029 127
XM_006724562.2 XP_006724625.1 11 - 12 1742 127
GenBank mRNA AK026254.1 - 2 - 3 303 127
AK026496.1 - 12 - 13 1883 127
AK291187.1 - 12 - 13 2170 127
AK292967.1 - 12 - 13 2013 127
BC031039.1 - 12 - 13 2024 127
CR749363.1 - 7 - 8 129

View Full Product Details