The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001309193.1 | NP_001296122.1 | 9 - 10 | 1909 | 75 |
NM_004442.7 | NP_004433.2 | 9 - 10 | 1912 | 75 | |
NM_017449.4 | NP_059145.2 | 9 - 10 | 1909 | 75 | |
XM_006710441.3 | XP_006710504.1 | 9 - 10 | 2005 | 75 | |
XM_006710442.3 | XP_006710505.1 | 9 - 10 | 1887 | 75 | |
GenBank mRNA | AB210018.1 | - | 9 - 10 | 1818 | 75 |
AF025304.1 | - | 9 - 10 | 1789 | 75 | |
BC007903.2 | - | 8 - 9 | 1687 | 75 | |
BC018763.1 | - | 7 - 8 | 1062 | 75 | |
D31661.1 | - | 9 - 10 | 1773 | 75 | |
D37827.1 | - | 3 - 4 | 240 | 75 | |
L41939.1 | - | 9 - 10 | 1871 | 75 |