The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145234.3 | NP_660277.2 | 1 - 2 | 167 | 92 |
XM_005262221.1 | XP_005262278.1 | 1 - 2 | 136 | 92 | |
XM_005262222.3 | XP_005262279.1 | 1 - 2 | 136 | 92 | |
XM_017029959.1 | XP_016885448.1 | 1 - 2 | 136 | 92 | |
GenBank mRNA | BC002909.2 | - | 1 - 2 | 84 | 92 |
BP377491.1 | - | 1 - 2 | 167 | 92 |