The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015245.2 | NP_056060.2 | 4 - 5 | 874 | 63 |
XM_005248964.3 | XP_005249021.1 | 5 - 6 | 925 | 63 | |
XM_006715036.2 | XP_006715099.1 | 5 - 6 | 925 | 63 | |
XM_011514431.2 | XP_011512733.1 | 5 - 6 | 925 | 63 | |
XM_011514432.2 | XP_011512734.1 | 5 - 6 | 925 | 63 | |
XM_011514433.1 | XP_011512735.1 | 5 - 6 | 925 | 63 | |
XM_011514434.2 | XP_011512736.1 | 4 - 5 | 862 | 63 | |
XM_011514435.2 | XP_011512737.1 | 4 - 5 | 862 | 63 | |
XM_011514436.2 | XP_011512738.1 | 5 - 6 | 4494 | 63 | |
GenBank mRNA | AK298987.1 | - | 4 - 5 | 832 | 63 |
BC022396.1 | - | 4 - 5 | 874 | 63 | |
BC031934.1 | - | 5 - 6 | 946 | 63 | |
BC132832.1 | - | 4 - 5 | 811 | 63 | |
D86982.1 | - | 4 - 5 | 871 | 63 |