Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLCO2B1
- Entrez Gene ID
-
11309
- Gene Name
- solute carrier organic anion transporter family member 2B1
- Gene Aliases
-
OATP-B, OATP2B1, OATPB, SLC21A9
- Chromosome Location
-
Chr.11: 75150987 - 75206400 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AL117465.1 |
- |
2 - 3 |
349 |
101 |
View Full Product Details