The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144703.2 | NP_653304.2 | 7 - 8 | 1192 | 97 |
XM_005260302.3 | XP_005260359.1 | 8 - 9 | 1304 | 97 | |
XM_011528605.2 | XP_011526907.1 | 9 - 10 | 1496 | 97 | |
XM_011528606.2 | XP_011526908.1 | 9 - 10 | 1414 | 97 | |
XM_011528607.2 | XP_011526909.1 | 8 - 9 | 1422 | 97 | |
XM_011528608.2 | XP_011526910.1 | 8 - 9 | 1385 | 97 | |
XM_011528609.2 | XP_011526911.1 | 7 - 8 | 1266 | 97 | |
XM_011528611.2 | XP_011526913.1 | 6 - 7 | 1110 | 97 | |
XM_011528612.2 | XP_011526914.1 | 9 - 10 | 1166 | 97 | |
XM_011528613.2 | XP_011526915.1 | 8 - 9 | 1026 | 97 | |
XM_011528614.2 | XP_011526916.1 | 8 - 9 | 1239 | 97 | |
XM_011528615.2 | XP_011526917.1 | 5 - 6 | 723 | 97 | |
XM_017027688.1 | XP_016883177.1 | 6 - 7 | 1099 | 97 | |
XM_017027689.1 | XP_016883178.1 | 6 - 7 | 1018 | 97 | |
XM_017027690.1 | XP_016883179.1 | 6 - 7 | 840 | 97 | |
GenBank mRNA | AK298748.1 | - | 7 - 8 | 901 | 97 |
BX370449.2 | - | 3 - 4 | 392 | 97 |