The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142326.1 | NP_001135798.1 | 11 - 12 | 1407 | 127 |
NM_001142327.1 | NP_001135799.1 | 12 - 13 | 1524 | 127 | |
NM_021145.3 | NP_066968.3 | 14 - 15 | 1761 | 127 | |
NR_024549.1 | 12 - 13 | 1735 | 127 | ||
NR_024550.1 | 12 - 13 | 1696 | 127 | ||
XM_011516735.2 | XP_011515037.1 | 12 - 13 | 2446 | 127 | |
XM_011516737.1 | XP_011515039.1 | 13 - 14 | 1571 | 127 | |
XM_011516739.2 | XP_011515041.1 | 11 - 12 | 1336 | 127 | |
XM_017012865.1 | XP_016868354.1 | 13 - 14 | 2540 | 127 | |
XM_017012866.1 | XP_016868355.1 | 11 - 12 | 2323 | 127 | |
XM_017012867.1 | XP_016868356.1 | 10 - 11 | 2991 | 127 | |
XM_017012868.1 | XP_016868357.1 | 13 - 14 | 1571 | 127 | |
XM_017012869.1 | XP_016868358.1 | 14 - 15 | 1600 | 127 | |
XM_017012870.1 | XP_016868359.1 | 11 - 12 | 1339 | 127 | |
XM_017012871.1 | XP_016868360.1 | 13 - 14 | 2712 | 127 | |
XM_017012872.1 | XP_016868361.1 | 12 - 13 | 2618 | 127 | |
XM_017012873.1 | XP_016868362.1 | 13 - 14 | 2507 | 127 | |
XM_017012874.1 | XP_016868363.1 | 13 - 14 | 1743 | 127 | |
XM_017012875.1 | XP_016868364.1 | 13 - 14 | 2546 | 127 | |
XM_017012876.1 | XP_016868365.1 | 11 - 12 | 1486 | 127 | |
XM_017012877.1 | XP_016868366.1 | 12 - 13 | 1605 | 127 | |
XM_017012878.1 | XP_016868367.1 | 14 - 15 | 2640 | 127 | |
GenBank mRNA | AF084530.1 | - | 12 - 13 | 1473 | 127 |
AF202144.1 | - | 12 - 13 | 1645 | 127 | |
AF202145.1 | - | 12 - 13 | 1684 | 127 | |
AK126664.1 | - | 12 - 13 | 2191 | 127 | |
AK296211.1 | - | 11 - 12 | 1300 | 127 | |
AK314622.1 | - | 12 - 13 | 1460 | 127 | |
BC007418.2 | - | 3 - 4 | 473 | 127 | |
BC007447.2 | - | 3 - 4 | 473 | 127 | |
BC070064.1 | - | 14 - 15 | 1707 | 127 |