The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021203.3 | NP_067026.3 | 3 - 4 | 536 | 68 |
GenBank mRNA | AF086914.1 | - | 2 - 3 | 274 | 68 |
AF141882.3 | - | 2 - 3 | 267 | 68 | |
AK027525.1 | - | 2 - 3 | 267 | 68 | |
AK075531.1 | - | 2 - 3 | 305 | 68 | |
BC063001.1 | - | 2 - 3 | 279 | 68 | |
BC065299.1 | - | 2 - 3 | 251 | 68 |