The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021203.3 | NP_067026.3 | 5 - 6 | 695 | 68 |
GenBank mRNA | AF086914.1 | - | 4 - 5 | 433 | 68 |
AF141882.3 | - | 4 - 5 | 426 | 68 | |
AK027307.1 | - | 1 - 2 | 938 | 68 | |
AK075531.1 | - | 4 - 5 | 464 | 68 | |
BC063001.1 | - | 4 - 5 | 438 | 68 | |
BC065299.1 | - | 4 - 5 | 410 | 68 |