The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000376.2 | NP_000367.1 | 9 - 10 | 1184 | 88 |
NM_001017535.1 | NP_001017535.1 | 10 - 11 | 1306 | 88 | |
NM_001017536.1 | NP_001017536.1 | 9 - 10 | 1575 | 88 | |
XM_006719587.3 | XP_006719650.1 | 8 - 9 | 1101 | 88 | |
XM_011538720.2 | XP_011537022.1 | 9 - 10 | 1216 | 88 | |
GenBank mRNA | AB307700.1 | - | 7 - 8 | 1024 | 88 |
AF026260.1 | - | 7 - 8 | 1024 | 88 | |
AK225844.1 | - | 9 - 10 | 1183 | 88 | |
AK299446.1 | - | 8 - 9 | 1248 | 88 | |
AK312267.1 | - | 9 - 10 | 1176 | 88 | |
BC033465.1 | - | 9 - 10 | 1163 | 88 | |
BC060832.1 | - | 9 - 10 | 1547 | 88 | |
HQ692836.1 | - | 7 - 8 | 1024 | 88 | |
J03258.1 | - | 9 - 10 | 1139 | 88 | |
X67482.1 | - | 8 - 9 | 1074 | 88 |