The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032236.5 | NP_115612.4 | 15 - 16 | 2202 | 157 |
XM_005246009.2 | XP_005246066.1 | 15 - 16 | 2206 | 157 | |
XM_006710951.2 | XP_006711014.2 | 15 - 16 | 2200 | 157 | |
XM_006710955.3 | XP_006711018.1 | 14 - 15 | 1984 | 157 | |
XM_011542262.2 | XP_011540564.1 | 15 - 16 | 2096 | 157 | |
XM_011542263.2 | XP_011540565.1 | 15 - 16 | 2096 | 157 | |
XM_011542264.1 | XP_011540566.1 | 14 - 15 | 1796 | 157 | |
XM_011542265.2 | XP_011540567.1 | 16 - 17 | 2762 | 157 | |
XM_011542267.2 | XP_011540569.1 | 15 - 16 | 2115 | 157 | |
XM_011542268.2 | XP_011540570.1 | 7 - 8 | 815 | 157 | |
XM_017002495.1 | XP_016857984.1 | 14 - 15 | 2005 | 157 | |
XM_017002496.1 | XP_016857985.1 | 15 - 16 | 2206 | 157 | |
XM_017002497.1 | XP_016857986.1 | 7 - 8 | 860 | 157 | |
GenBank mRNA | AF502942.1 | - | 15 - 16 | 2104 | 157 |
AK021830.1 | - | 4 - 5 | 1535 | 157 | |
AK026707.1 | - | 4 - 5 | 866 | 157 | |
AK226150.1 | - | 14 - 15 | 2000 | 157 | |
AK315005.1 | - | 15 - 16 | 1982 | 157 | |
BC007326.2 | - | 6 - 7 | 770 | 157 | |
BC104896.1 | - | 15 - 16 | 1998 | 157 | |
BC143343.1 | - | 15 - 16 | 1998 | 157 | |
BC143349.1 | - | 15 - 16 | 1998 | 157 |