The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032730.1 | NP_001027902.1 | 5 - 6 | 904 | 136 |
NM_032236.5 | NP_115612.4 | 5 - 6 | 904 | 136 | |
XM_005246009.2 | XP_005246066.1 | 5 - 6 | 908 | 136 | |
XM_006710951.2 | XP_006711014.2 | 5 - 6 | 908 | 136 | |
XM_006710955.3 | XP_006711018.1 | 5 - 6 | 884 | 136 | |
XM_011542262.2 | XP_011540564.1 | 5 - 6 | 801 | 136 | |
XM_011542263.2 | XP_011540565.1 | 5 - 6 | 801 | 136 | |
XM_011542264.1 | XP_011540566.1 | 4 - 5 | 498 | 136 | |
XM_011542265.2 | XP_011540567.1 | 6 - 7 | 1464 | 136 | |
XM_011542267.2 | XP_011540569.1 | 5 - 6 | 801 | 136 | |
XM_017002495.1 | XP_016857984.1 | 5 - 6 | 908 | 136 | |
XM_017002496.1 | XP_016857985.1 | 5 - 6 | 908 | 136 | |
GenBank mRNA | AF502942.1 | - | 5 - 6 | 806 | 136 |
AK074247.1 | - | 5 - 6 | 801 | 136 | |
AK226150.1 | - | 5 - 6 | 900 | 136 | |
AK315005.1 | - | 5 - 6 | 684 | 136 | |
BC067261.1 | - | 5 - 6 | 872 | 136 | |
BC103724.1 | - | 5 - 6 | 846 | 136 | |
BC104896.1 | - | 5 - 6 | 700 | 136 | |
BC143343.1 | - | 5 - 6 | 700 | 136 | |
BC143349.1 | - | 5 - 6 | 700 | 136 | |
BC143354.1 | - | 5 - 6 | 700 | 136 |