The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024551.2 | NP_078827.2 | 2 - 3 | 369 | 71 |
XM_005253789.1 | XP_005253846.1 | 2 - 3 | 491 | 71 | |
XM_006719018.2 | XP_006719081.1 | 3 - 4 | 488 | 71 | |
XM_011521024.2 | XP_011519326.1 | 3 - 4 | 499 | 71 | |
GenBank mRNA | AK127196.1 | - | 3 - 4 | 424 | 71 |
AK128511.1 | - | 2 - 3 | 423 | 71 | |
AY424280.1 | - | 1 - 2 | 248 | 71 | |
BC051858.2 | - | 2 - 3 | 369 | 71 |